Learn More
Aviva Systems Biology Corp KDM7A Antibody - C region (ARP58206_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58206_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JHDM1D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human JHDM1D. Alternate names: KIAA1718, KDM7AApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53763