Learn More
Creative Bioarray JHOS-4 Cells, Human, Serous Cyst Adenocarcinoma, 44 Y, Female, Metastatic Peritoneum, Doubling Time 70 hours, Storage Liquid Nitrogen, Shipping Dry Ice, Available Omics Data: Genomics, Proteomics, Transcriptomics
Catalog No. NC3444016
Encompass
JHOS-4 Cells, Human, Serous Cyst Adenocarcinoma, 44 Y, Female, Metastatic Peritoneum, Doubling Time 70 hours, Storage Liquid Nitrogen, Shipping Dry Ice, Available Omics Data: Genomics, Proteomics, Transcriptomics
Catalog No. NC3444016 Supplier Creative Bioarray Supplier No. CSCC6613J
JHOS-4 Cells, Human, Serous Cyst Adenocarcinoma, 44 Y, Female, Metastatic Peritoneum, Doubling Time 70 hours, Storage Liquid Nitrogen, Shipping Dry Ice, Available Omics Data: Genomics, Proteomics, Transcriptomics