Learn More
Medchemexpress LLC HY-122218 5mg , JHU-083 CAS:1998725-11-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1222185MG
Encompass_Preferred
HY-122218 5mg JHU-083 CAS: 1998725-11-3 JHU-083, a prodrug of 6-diazo-5-oxo-L-norleucine (DON; HY-108357), is an orally active and selective glutaminase antagonist. JHU-083 blocks glutaminase activity in brain CD11b+ cells and experimental cerebral malaria (ECM) resulting in a net decrease of glutamate levels in the animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9651