Learn More
Selleck Chemical LLC JHU-083 S8891-25MG
SDP
Catalog No. 5000025374
Encompass_Preferred
JHU-083 (Ethyl 2-(2-Amino-4-methylpentanamido)-DON) is a novel prodrug of DON JHU-083 selectively blocks glutaminase activity in brain CD11b cells and prevents depression-associated behaviors induced by chronic social defeat stress Solutions are unstable and should be fresh-prepared
Catalog No. 50-000-25374 Supplier Selleck Chemical LLC Supplier No. S889125MG
JHU-083 (Ethyl 2-(2-Amino-4-methylpentanamido)-DON) is a novel prodrug of DON JHU-083 selectively blocks glutaminase activity in brain CD11b cells and prevents depression-associated behaviors induced by chronic social defeat stress Solutions are unstable and should be fresh-prepared