Learn More
TARGETMOL CHEMICALS INC JHU 75528 5MG
SDP
Catalog No. 502579128
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. JHU 75528 (JHU-75528) is a novel PET tracer with inhibitory effects on CB that can be used to study the cannabinoid system in schizophrenic patients. purity: 99%
Catalog No. 50-257-9128 Supplier TARGETMOL CHEMICALS INC Supplier No. T776345MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. JHU 75528 (JHU-75528) is a novel PET tracer with inhibitory effects on CB that can be used to study the cannabinoid system in schizophrenic patients. purity: 99%