Learn More
TARGETMOL CHEMICALS INC JHU395 5MG
SDP
Catalog No. 502581516
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. JHU395 is an orally-bioavailable prodrug of a lipophilic glutamine antagonist (GA) 6-diazo-5-oxo-L-norleucine (DON). It demonstrates delivery of DON to malignant peripheral nerve sheath tumor (MPNST) both in vitro and in vivo resulting in significant antitumor activity in MPNST. Additionally JHU395 exhibits plasma stability. purity: 99%
Catalog No. 50-258-1516 Supplier TARGETMOL CHEMICALS INC Supplier No. T117175MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. JHU395 is an orally-bioavailable prodrug of a lipophilic glutamine antagonist (GA) 6-diazo-5-oxo-L-norleucine (DON). It demonstrates delivery of DON to malignant peripheral nerve sheath tumor (MPNST) both in vitro and in vivo resulting in significant antitumor activity in MPNST. Additionally JHU395 exhibits plasma stability. purity: 99%