Learn More
Medchemexpress LLC Ji-101 | 900573-88-8 | MFCD26142530 | 99.8% | 466.33 | C22H20BrN5O2 | 10 MG
SDP
Catalog No. 5000288706
Encompass_Preferred
JI-101 is an orally available multi-kinase inhibitor that targets VEGFR2, PDGFRβ, and EphB4 and has demonstrated potent anti-cancer activity. It is provided as a white to off-white solid powder for research use and is commonly supplied in a 10 mg pack.
  • Orally available multi-kinase inhibitor active against VEGFR2, PDGFRβ, and EphB4.
  • High purity (99.78%).
  • Molecular formula C22H20BrN5O2; molecular weight 466.33 g/mol.
  • White to off-white solid, soluble in DMSO at ≥ 100 mg/mL.
  • Intended for research use only; not for human administration.
Catalog No. 50-002-88706 Supplier Medchemexpress LLC Supplier No. HY1626510MG
JI-101 is an orally available multi-kinase inhibitor that targets VEGFR2, PDGFRβ, and EphB4 and has demonstrated potent anti-cancer activity. It is provided as a white to off-white solid powder for research use and is commonly supplied in a 10 mg pack.
  • Orally available multi-kinase inhibitor active against VEGFR2, PDGFRβ, and EphB4.
  • High purity (99.78%).
  • Molecular formula C22H20BrN5O2; molecular weight 466.33 g/mol.
  • White to off-white solid, soluble in DMSO at ≥ 100 mg/mL.
  • Intended for research use only; not for human administration.