Learn More
Medchemexpress LLC HY-117113 10mg Medchemexpress, JI051 CAS:2234281-75-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11711310MG
Encompass_Preferred
Medchemexpress, HY-117113 10mg JI051 CAS:2234281-75-3 JI051 is a stabilizer for the Hes1-PHB2 interaction, interacts with a cancer-associated protein chaperone prohibitin 2 (PHB2), induces cell-cycle arrest by inhibiting the Notch downstream effector gene Hes1. Anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9454