Learn More
Medchemexpress LLC HY-117113 5mg Medchemexpress, JI051 CAS:2234281-75-3 Purity:>98%
SDP
Catalog No. 501932516
Encompass_Preferred
Medchemexpress, HY-117113 5mg JI051 CAS:2234281-75-3 JI051 is a stabilizer for the Hes1-PHB2 interaction, interacts with a cancer-associated protein chaperone prohibitin 2 ( PHB2 ), induces cell-cycle arrest by inhibiting the Notch downstream effector gene Hes1. Anti-cancer activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2516 Supplier Medchemexpress LLC Supplier No. HY1171135MG
Medchemexpress, HY-117113 5mg JI051 CAS:2234281-75-3 JI051 is a stabilizer for the Hes1-PHB2 interaction, interacts with a cancer-associated protein chaperone prohibitin 2 ( PHB2 ), induces cell-cycle arrest by inhibiting the Notch downstream effector gene Hes1. Anti-cancer activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.