Learn More
Apexbio Technology LLC JIB-04 199596-05-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063793
Encompass_Preferred
JIB-04 (CAS 199596-05-9) is a selective small-molecule inhibitor targeting Jumonji histone demethylases specifically suppressing enzymes including JARID1A JMJD2 family members (JMJD2A-E) and JMJD3 Structurally categorized as a pyridine hydrazone derivative JIB-04 inhibits demethylase activity independently of competitive binding with -ketoglutarate In vitro assays revealed IC50 values ranging from 230 nM to 1100 nM across various Jumonji enzymes Additionally JIB-04 demonstrates preferential inhibition of cancer cell proliferation versus normal cells notably in lung and prostate cancer cell lines and effectively reduces tumor growth in mouse xenograft models highlighting its relevance for cancer research
Catalog No. 50-000-63793 Supplier Apexbio Technology LLC Supplier No. B15795.1
JIB-04 (CAS 199596-05-9) is a selective small-molecule inhibitor targeting Jumonji histone demethylases specifically suppressing enzymes including JARID1A JMJD2 family members (JMJD2A-E) and JMJD3 Structurally categorized as a pyridine hydrazone derivative JIB-04 inhibits demethylase activity independently of competitive binding with -ketoglutarate In vitro assays revealed IC50 values ranging from 230 nM to 1100 nM across various Jumonji enzymes Additionally JIB-04 demonstrates preferential inhibition of cancer cell proliferation versus normal cells notably in lung and prostate cancer cell lines and effectively reduces tumor growth in mouse xenograft models highlighting its relevance for cancer research