Learn More
Apexbio Technology LLC JIB-04, 50mg. Cas: 199596-05-9 MFCD: MFCD26936344. Jumonji histone demethylase inihibitor
SDP
Supplier:  Apexbio Technology LLC B157950
Encompass_Preferred
JIB-04 is a small molecule inhibitor of Jumonji histone demethylase with IC50 values of [1].JIB-04 is a pyridine hydrazone. It is a pan -selective inhibitor of Jumonji histone demethylases. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5265