Learn More
Cole-Parmer Jiffy-Jack Lab Jack, 10" x 10" Top, 186 Lb Capacity
Catalog No. 50632399 Shop All Cole-Parmer Products
Encompass_Preferred
These laboratory scissor jacks provide stable, height-adjustable support for hot plates, baths, glassware, and many other items. Positioners are constructed of rugged, chemical-resistant anodized aluminum with stainless steel operating screw. Top-plate skid pad prevents slipping. Each positioner is precision engineered for smooth, continuous operation throughout its range. Easily set the exact height needed with the turn of a knob.
Catalog No. 50-632-399 Supplier Cole-Parmer Supplier No. 0805740
These laboratory scissor jacks provide stable, height-adjustable support for hot plates, baths, glassware, and many other items. Positioners are constructed of rugged, chemical-resistant anodized aluminum with stainless steel operating screw. Top-plate skid pad prevents slipping. Each positioner is precision engineered for smooth, continuous operation throughout its range. Easily set the exact height needed with the turn of a knob.