Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boekel Scientific™ Jitterbug 4 Incubated Microplate Shaker
SDP

Catalog No. 04100565 Shop All Boekel Scientific Products
Change view
Click to view available options
Holds:
4 standard microplates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
04-100-565 4 standard microplates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-100-565 Supplier Boekel Scientific™ Supplier No. 270440
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Jitterbug 4 Plate heated microplate shaker is ideal for immunoassays like ELISA, PCR and other protocols requiring heating and shaking of 4 standard microplates or strip trays.

  • Temperature Range: Ambient +5°C to 60°C
  • Temperature Stability: ±0.1°C
  • Temperature Uniformity: ±0.2°C
  • Temperature Display Resolution: 0.1°C
  • Heating Rate: Ambient to 37°C in 15 minutes
  • Overall Dimensions (WxDxH): 15 x 15.5 x 5.5 in.
  • Orbit Diameter: 2 mm
  • Mixing Speed: 250 to 1200 RPM
  • Timer Maximum Setpoint: 96 hours 59 minutes
  • Timer Resolution: 1 minute
  • Shipping Weight: 22 lbs
  • Electrical: 100-240VAC 12VDC

Specifications

Orbit 2 mm
Load Bearing Capacity (Metric) 0.5 kg
Speed Range 250 to 1200 rpm
Voltage 100/240 VAC
Temperature Range (Metric) Ambient +5°C to 60°C
Amperage 5.0A (12V DC)
Rotation Orbital
Holds 4 standard microplates
Product Type Shaking Incubator
Plug Type 100-240VAC, 12VDC adapter
Material ABS
Length (English) 15.5 in.
Length (Metric) 39.4 cm
Accuracy ±0.3°C
Model Jitterbug 4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.