Learn More
Medchemexpress LLC HY-108613 5mg Medchemexpress, JJKK 048 CAS:1515855-97-6 Purity:>98%
SDP
Catalog No. 501968597
Encompass_Preferred
Medchemexpress, HY-108613 5mg JJKK 048 CAS:1515855-97-6 JJKK 048 is an ultrapotent and highly selective inhibitor of Monoacylglycerol lipase (MAGL). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8597 Supplier Medchemexpress LLC Supplier No. HY1086135MG
Medchemexpress, HY-108613 5mg JJKK 048 CAS:1515855-97-6 JJKK 048 is an ultrapotent and highly selective inhibitor of Monoacylglycerol lipase (MAGL). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.