Learn More
Enzo Life Sciences JK 184 (5mg). CAS: 315703-52-7
SDP
Catalog No. 502013419
Encompass_Preferred
Alternative name: N-(4-ethoxyphenyl)-4-(2-methylimidazo[1,2-a]pyridin-3-yl)-1,3-thiazol-2-amine, Hh signaling antagonist VII. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.
Catalog No. 50-201-3419 Supplier Enzo Life Sciences Supplier No. ENZCHM1290005
Alternative name: N-(4-ethoxyphenyl)-4-(2-methylimidazo[1,2-a]pyridin-3-yl)-1,3-thiazol-2-amine, Hh signaling antagonist VII. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.