Learn More
Enzo Life Sciences ET A receptor antagonist [JKC-301] (1mg)
SDP
Supplier:  Enzo Life Sciences ALX155012P001
Encompass_Preferred
Very potent ETA antagonist. Formula: C32H44N6O7. MW: 624.7. Purity: ≥96%. Appearance: White to off-white lyophilized powder. Solubility: 1mg/ml completely soluble in 5% acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-7803