Learn More
Across International JKEM DVR-200 DVR-300 Digital Vacuum Regulator
SDP
Catalog No. NC1814830
Encompass_Preferred
JKEM model DVR-200, an ideal instrument for laboratory vacuum regulation.Designed for continuous vacuum sources, such as oil-filled vacuum pumps, aspirators, or systems that are always on, like in-house vacuum systems. The DVR-200 regulates pressure by opening and closing a valve that separates the vacuum source from the system being evacuated. In most cases, the DVR-200 is the best choice.
Catalog No. NC1814830 Supplier Across International Supplier No. DVR200
JKEM model DVR-200, an ideal instrument for laboratory vacuum regulation.Designed for continuous vacuum sources, such as oil-filled vacuum pumps, aspirators, or systems that are always on, like in-house vacuum systems. The DVR-200 regulates pressure by opening and closing a valve that separates the vacuum source from the system being evacuated. In most cases, the DVR-200 is the best choice.