Learn More
J Kem Scientific Inc J-Kem Scientific Gemini-T-S, Dual-Channel Temperature Controller, 0.1°C Regulation Accuracy, 1800 Watts Max Power
SDP
Catalog No. NC1862848
Encompass
The J-Kem Scientific Gemini-T-S is a dual-channel temperature controller that delivers precise temperature regulation between -200 to 250°C, making it ideal for laboratory applications. With features like advanced microprocessor PID control, a 100-hour timer, and robust safety mechanisms, it stands out for reliability. Each channel outputs 1200 watts, supporting demanding tasks with ease. The device also integrates a USB port for enhanced data management, providing comprehensive control and efficiency in experiments.
Catalog No. NC1862848 Supplier J Kem Scientific Inc Supplier No. GEMINITS
The J-Kem Scientific Gemini-T-S is a dual-channel temperature controller that delivers precise temperature regulation between -200 to 250°C, making it ideal for laboratory applications. With features like advanced microprocessor PID control, a 100-hour timer, and robust safety mechanisms, it stands out for reliability. Each channel outputs 1200 watts, supporting demanding tasks with ease. The device also integrates a USB port for enhanced data management, providing comprehensive control and efficiency in experiments.