Learn More
Ace Glass, Inc. Short path still body, vacuum jacketed with Vigreux indents, 24/40 path joints, 10/30 thermometer joint
SDP
Catalog No. 50973253
Encompass_Preferred
Short path still body, vacuum jacketed with Vigreux indents, 24/40 path joints, 10/30 thermometer joint
Catalog No. 50-973-253 Supplier Ace Glass, Inc. Supplier No. 655408
Short path still body, vacuum jacketed with Vigreux indents, 24/40 path joints, 10/30 thermometer joint