Learn More
Ace Glass, Inc. Short path still, 500mL, complete with distillation flask, gas inlet tube with adapter, vacuum jacketed still body with Vigreux indents, thermometer, cow receiver and (3)250mLreceiving flasks
SDP
Catalog No. 50973255
Encompass_Preferred
24/40 jointed, jacked short path still with indents. Extension tube from still body allows positioning of drop directly over receiver flask. Gas inlet tube is positioned in the distillation flask with an ACE 5028 adapter and secured with a #7 Ace-Thred. Thermometer joint is 10/30. Complete system consists of 500mL distillation flask, still body, cow receiver, gas inlet tube, threaded adapter, 3 250mL receiver flasks and a thermometer.
Catalog No. 50-973-255 Supplier Ace Glass, Inc. Supplier No. 655450
24/40 jointed, jacked short path still with indents. Extension tube from still body allows positioning of drop directly over receiver flask. Gas inlet tube is positioned in the distillation flask with an ACE 5028 adapter and secured with a #7 Ace-Thred. Thermometer joint is 10/30. Complete system consists of 500mL distillation flask, still body, cow receiver, gas inlet tube, threaded adapter, 3 250mL receiver flasks and a thermometer.