Learn More
Beckman Coulter JLA-16.250 Fixed-Angle Aluminum Rotor, Biosafety Lid - 6 x 250 mL, 16,000 rpm, 38,400 x g
Catalog No. NC1887408
Encompass
JLA-16.250 Fixed-Angle Rotor with Biosafety Lid, Aluminum. Major applications: General purpose, large-volume and multitube processing. Lightweight alternative to conventional 250 rotors; empty rotor weight is 10.3 kg (22.7 lb). For use in Avanti J Series and J2 Series centrifuges. The maximum speed is 14,000 rpm in J2 Series centrifuges.
This item is not returnable. View return policy
Catalog No. NC1887408 Supplier Beckman Coulter Supplier No. 363930
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
JLA-16.250 Fixed-Angle Rotor with Biosafety Lid, Aluminum. Major applications: General purpose, large-volume and multitube processing. Lightweight alternative to conventional 250 rotors; empty rotor weight is 10.3 kg (22.7 lb). For use in Avanti J Series and J2 Series centrifuges. The maximum speed is 14,000 rpm in J2 Series centrifuges.