Learn More
Beckman Coulter J-LITE JLA-8.1000 Fixed-Angle Rotor for Avanti J-26 Series, 6 x 1,000 mL (Beckman Coulter, MPN 363688)
Catalog No. NC2131297
Encompass
The J-LITE JLA-8.1000 fixed-angle aluminum rotor (MPN 363688) is engineered for high-capacity, general-purpose centrifugation and large-volume pelleting applications such as bacteria, cell organelles, viruses, and precipitates. It accepts six 1,000 mL bottles and is rated for demanding laboratory processing.
  • Accommodates six 1,000 mL bottles for large-volume processing.
  • Rated to 8,000 rpm, producing up to 15,970 x g for efficient pelleting.
  • Anodized aluminum construction for durable, lightweight performance.
  • 20° tube angle to optimize sedimentation and pellet formation.
  • Lightweight design with an empty weight of approximately 16.8 kg for easier handling.
  • Includes bottle rack, spatula, and basic tool kit for handling and maintenance.
  • Compatible with a range of adapters to accept different bottle types.
This item is not returnable. View return policy
Catalog No. NC2131297 Supplier Beckman Coulter Supplier No. 363688
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The J-LITE JLA-8.1000 fixed-angle aluminum rotor (MPN 363688) is engineered for high-capacity, general-purpose centrifugation and large-volume pelleting applications such as bacteria, cell organelles, viruses, and precipitates. It accepts six 1,000 mL bottles and is rated for demanding laboratory processing.
  • Accommodates six 1,000 mL bottles for large-volume processing.
  • Rated to 8,000 rpm, producing up to 15,970 x g for efficient pelleting.
  • Anodized aluminum construction for durable, lightweight performance.
  • 20° tube angle to optimize sedimentation and pellet formation.
  • Lightweight design with an empty weight of approximately 16.8 kg for easier handling.
  • Includes bottle rack, spatula, and basic tool kit for handling and maintenance.
  • Compatible with a range of adapters to accept different bottle types.