Learn More
Beckman Coulter J-LITE JLA-16.250 Fixed-Angle Rotor, 6 x 250 mL, 16,000 rpm
Catalog No. NC2210599
Encompass
The J-LITE JLA-16.250 is a fixed-angle aluminum rotor designed for large-volume, high-speed centrifugation. It holds six 250 mL bottles (1,500 mL nominal capacity), is rated to 16,000 rpm (38,400 x g), and features a single-lock lid for secure operation. Suitable for general-purpose and multitube processing, and compatible with Avanti J Series and J2 Series centrifuges.
  • Holds six 250 mL bottles (1,500 mL nominal capacity).
  • Anodized aluminum construction.
  • Rated to 16,000 rpm (38,400 x g); limited to 14,000 rpm in certain centrifuge models.
  • Empty rotor weight approximately 10.3 kg (22.7 lb).
  • Single-lock lid for secure containment.
  • Intended for general-purpose, large-volume, multitube processing.
Catalog No. NC2210599 Supplier Beckman Coulter Supplier No. 363934
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The J-LITE JLA-16.250 is a fixed-angle aluminum rotor designed for large-volume, high-speed centrifugation. It holds six 250 mL bottles (1,500 mL nominal capacity), is rated to 16,000 rpm (38,400 x g), and features a single-lock lid for secure operation. Suitable for general-purpose and multitube processing, and compatible with Avanti J Series and J2 Series centrifuges.
  • Holds six 250 mL bottles (1,500 mL nominal capacity).
  • Anodized aluminum construction.
  • Rated to 16,000 rpm (38,400 x g); limited to 14,000 rpm in certain centrifuge models.
  • Empty rotor weight approximately 10.3 kg (22.7 lb).
  • Single-lock lid for secure containment.
  • Intended for general-purpose, large-volume, multitube processing.