Learn More
Agilent Technologies JM101 COMPETENT CELLS
Catalog No. 50125051
Encompass
JM101 is a recombination- and EcoK restriction-proficient (RecA +, Hsdr + ) host strain. JM101 cells contain the lacIqZDeltaM15 gene on the F episome, allowing blue-white screening for recombinant plasmids.
Catalog No. 50-125-051 Supplier Agilent Technologies Supplier No. 200234
JM101 is a recombination- and EcoK restriction-proficient (RecA +, Hsdr + ) host strain. JM101 cells contain the lacIqZDeltaM15 gene on the F episome, allowing blue-white screening for recombinant plasmids.