Learn More
INTACT GENOMICS INC JM109 Chemically Competent Cells 6x200µl
Catalog No. 5000082911
Encompass
Intact Genomics JM109 Chemically Competent E. coli cells are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning.ig™ JM109 chemically competent cells have the following features: • recA– and endA– to minimize recombination and improve the quality of plasmid DNA. • Contains F’ episome for blue/white screening.Product Components & Storage:1) ig™ JM109 competent cells: -80 ºC2) pUC19 control DNA: -20 ºC3) Recovery medium: 4 ºC
Catalog No. 50-000-82911 Supplier INTACT GENOMICS INC Supplier No. 106224
Intact Genomics JM109 Chemically Competent E. coli cells are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning.ig™ JM109 chemically competent cells have the following features: • recA– and endA– to minimize recombination and improve the quality of plasmid DNA. • Contains F’ episome for blue/white screening.Product Components & Storage:1) ig™ JM109 competent cells: -80 ºC2) pUC19 control DNA: -20 ºC3) Recovery medium: 4 ºC