Learn More
Agilent Technologies JM109 COMPETENT CELLS
Catalog No. 50125052
Encompass
The JM109 strain is recA-, endA- and is appropriate for routine cloning applications. JM109 cells containthe lacIqZDeltaM15 gene on the F episome, allowing blue-white screening for recombinant plasmids.
Catalog No. 50-125-052 Supplier Agilent Technologies Supplier No. 200235
The JM109 strain is recA-, endA- and is appropriate for routine cloning applications. JM109 cells containthe lacIqZDeltaM15 gene on the F episome, allowing blue-white screening for recombinant plasmids.