Learn More
Agilent Technologies JM110 COMPETENT CELLS
Catalog No. 50125055
Encompass
JM110 Competent Cells are useful for preparing plasmid or phagemid DNA that is free of Dam or Dcm methylation so that the DNA can be restriction digested by one or more methylation-sensitive restriction enzymes.
Catalog No. 50-125-055 Supplier Agilent Technologies Supplier No. 200239
JM110 Competent Cells are useful for preparing plasmid or phagemid DNA that is free of Dam or Dcm methylation so that the DNA can be restriction digested by one or more methylation-sensitive restriction enzymes.