Learn More
Uline Jumbo Heavy Duty Storage Cabinet
Catalog No. NC2587860
Encompass
Jumbo Heavy Duty Storage Cabinet - 48 x 18 x 78", Assembled, Gray, heavy-duty steel construction, locking doors, 4 adjustable shelves (400lb capacity each)
Catalog No. NC2587860 Supplier Uline Supplier No. H3617AGR
Jumbo Heavy Duty Storage Cabinet - 48 x 18 x 78", Assembled, Gray, heavy-duty steel construction, locking doors, 4 adjustable shelves (400lb capacity each)