Learn More
Aviva Systems Biology Corp JMJD1B Antibody - C-terminal region (ARP39268_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39268_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against JMJD1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human JMJD1B. Alternate names: 5qNCA, NET22, C5orf7, JMJD1BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56962