Learn More
Medchemexpress LLC JMJD1C-IN-1 | 861224-48-8 | 100.0% | 317.36 | 100 MG
SDP
Catalog No. 5000392727
Encompass_Preferred
This product is an orally active and selective inhibitor that targets JMJD1C, preventing its binding to H3K9me2 peptide substrate. It improves antitumor immunity by disrupting intratumoral regulatory T cell fitness, promoting H3K9me2 accumulation to downregulate PD1 expression, and enhancing STAT3 activation. It has shown dose-dependent antitumor efficacy in multiple mouse tumor models and is valuable for tumor immunotherapy research.
  • Orally active and selective inhibition of target.
  • Inhibits target binding to H3K9me2 peptide substrate.
  • Disrupts intratumoral regulatory T cell fitness for improved antitumor immunity.
  • Promotes H3K9me2 accumulation, downregulating PD1 expression.
  • Enhances STAT3 activation by reducing demethylation.
  • Demonstrates dose-dependent antitumor efficacy across various mouse tumor models.
  • Exhibited no significant toxicity in animal models.
Catalog No. 50-003-92727 Supplier Medchemexpress LLC Supplier No. HY162275100MG
This product is an orally active and selective inhibitor that targets JMJD1C, preventing its binding to H3K9me2 peptide substrate. It improves antitumor immunity by disrupting intratumoral regulatory T cell fitness, promoting H3K9me2 accumulation to downregulate PD1 expression, and enhancing STAT3 activation. It has shown dose-dependent antitumor efficacy in multiple mouse tumor models and is valuable for tumor immunotherapy research.
  • Orally active and selective inhibition of target.
  • Inhibits target binding to H3K9me2 peptide substrate.
  • Disrupts intratumoral regulatory T cell fitness for improved antitumor immunity.
  • Promotes H3K9me2 accumulation, downregulating PD1 expression.
  • Enhances STAT3 activation by reducing demethylation.
  • Demonstrates dose-dependent antitumor efficacy across various mouse tumor models.
  • Exhibited no significant toxicity in animal models.