Learn More
Medchemexpress LLC JMJD1C-IN-1 | 861224-48-8 | 99.99% | 317.36 | 25 MG
SDP
Catalog No. 5000392635
Encompass_Preferred
JMJD1C-IN-1 is an orally active and selective inhibitor of JMJD1C (IC50 = 0.59 μM, Kd = 1.96 μM). It inhibits the binding of JMJD1C to H3K9me2 peptide substrate and disrupts intratumoral regulatory T (Treg) cell fitness by promoting H3K9me2 accumulation to downregulate PD1 expression and reducing STAT3 demethylation to enhance STAT3 activation. It demonstrates dose-dependent antitumor efficacy in multiple mouse tumor models and can be used for the study of tumor immunotherapy by selectively targeting intratumoral Treg cells.
  • Orally active and selective inhibitor of JMJD1C.
  • Disrupts intratumoral regulatory T (Treg) cell fitness by dual mechanisms.
  • Demonstrates dose-dependent antitumor efficacy in multiple mouse tumor models.
  • Can be used for the study of tumor immunotherapy by selectively targeting intratumoral Treg cells.
Catalog No. 50-003-92635 Supplier Medchemexpress LLC Supplier No. 25MGHY162275
JMJD1C-IN-1 is an orally active and selective inhibitor of JMJD1C (IC50 = 0.59 μM, Kd = 1.96 μM). It inhibits the binding of JMJD1C to H3K9me2 peptide substrate and disrupts intratumoral regulatory T (Treg) cell fitness by promoting H3K9me2 accumulation to downregulate PD1 expression and reducing STAT3 demethylation to enhance STAT3 activation. It demonstrates dose-dependent antitumor efficacy in multiple mouse tumor models and can be used for the study of tumor immunotherapy by selectively targeting intratumoral Treg cells.
  • Orally active and selective inhibitor of JMJD1C.
  • Disrupts intratumoral regulatory T (Treg) cell fitness by dual mechanisms.
  • Demonstrates dose-dependent antitumor efficacy in multiple mouse tumor models.
  • Can be used for the study of tumor immunotherapy by selectively targeting intratumoral Treg cells.