Learn More
Epigentek Inc Epigenase JMJD2 Demethylase Activity/Inhibition Assay Kit (Fluorometric) (96 assays)
SDP
Catalog No. 501096318
Encompass_Preferred
The Epigenase JMJD2 Demethylase Activity/Inhibition Assay Kit (Fluorometric) is a complete set of optimized reagents, designed for an easy and fast fluorometric measurement of JMJD2 activity or inhibition. The antibody-based, immunospecific method directly detects JMJD2-converted demethylated products, rather than by- products, in a 96 stripwell microplate format.
Catalog No. 50-109-6318 Supplier Epigentek Inc Supplier No. P308196
The Epigenase JMJD2 Demethylase Activity/Inhibition Assay Kit (Fluorometric) is a complete set of optimized reagents, designed for an easy and fast fluorometric measurement of JMJD2 activity or inhibition. The antibody-based, immunospecific method directly detects JMJD2-converted demethylated products, rather than by- products, in a 96 stripwell microplate format.