Learn More
Cayman Chemical JMJD2A Tudor DomaIns PosItIve
SDP
Catalog No. 502892342
Encompass_Preferred
INDIGOs assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to AVPR1B Reporter Cells provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples the reference activator Vasopressin Luciferase Detection Reagents and a cell culture-ready assay plate.
Catalog No. 50-289-2342 Supplier Cayman Chemical Supplier No. 70096512.5 NMOL
INDIGOs assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to AVPR1B Reporter Cells provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples the reference activator Vasopressin Luciferase Detection Reagents and a cell culture-ready assay plate.