Learn More
Cayman Chemical JMJD2A tudor domaInshumn100ug
SDP
Catalog No. 502952892
Encompass_Preferred
Source: Expressed in E. coli • Amino acids: AA: 888-1,023 (N-terminal truncation) • MW: 42.2 kDa
Catalog No. 50-295-2892 Supplier Cayman Chemical Supplier No. 14134100 UG
Source: Expressed in E. coli • Amino acids: AA: 888-1,023 (N-terminal truncation) • MW: 42.2 kDa