Learn More
Aviva Systems Biology Corp JMJD2B Antibody - middle region (ARP58207_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58207_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JMJD2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human JMJD2B. Alternate names: FLJ44906, KDM4B, KIAA0876, JMJD2BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62258