Learn More
Aviva Systems Biology Corp JMJD2C Antibody - C-terminal region (ARP32974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JMJD2C. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human JMJD2C. Alternate names: FLJ25949, GASC1, JHDM3C, JMJD2C, KIAA0780, bA146B14.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52881