Learn More
Cayman Chemical JMJD2C tudor domaInhumnr 100UG
SDP
Catalog No. 502892974
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • Amino acids: 869-1,003 (tudor domain) • Mr: 39.89 kDa
Catalog No. 50-289-2974 Supplier Cayman Chemical Supplier No. 9001953100 UG
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • Amino acids: 869-1,003 (tudor domain) • Mr: 39.89 kDa