Learn More
Eurofins Discoverx JMJD2D, 50 µg. (EPI032)
Supplier:  Eurofins Discoverx EPI032
Encompass
JMJD2D is purified, N terminal 6xHis tagged recombinant JMJD2D enzyme. This enzyme contains amino acids 1-336. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5254