Learn More
Aviva Systems Biology Corp JMJD2D Antibody - C-terminal region (ARP35946_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35946_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against JMJD2D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human JMJD2D. Alternate names: JMJD2DApplicationWB. Species reactivity: Human
Catalog No. 50-014-55937