Learn More
Aviva Systems Biology Corp JMJD2D Antibody - C-terminal region (ARP39325_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39325_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JMJD2D. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human JMJD2D. Alternate names: FLJ10251, JMJD2DApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-56986