Learn More
Eurofins Discoverx JMJD2E, 50 µg. (EPI033)
Supplier:  Eurofins Discoverx EPI033
Encompass
JMJD2E is purified, N terminal 6xHis tagged recombinant JMJD2E enzyme. This enzyme contains amino acids 1141-1641. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5257