Learn More
Eurofins Discoverx JMJD3, 50 µg. (EPI034)
Supplier:  Eurofins Discoverx EPI034
Encompass
JMJD3 is purified, untagged recombinant JMJD3 enzyme. This enzyme contains amino acids 1810-2120. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5258