Learn More
Aviva Systems Biology Corp JMJD3 Antibody - middle region (ARP40102_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40102_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against JMJD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human JMJD3. Alternate names: KIAA0346, JMJD3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53174