Learn More
Aviva Systems Biology Corp JMJD4 Antibody - N-terminal region (ARP39572_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39572_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JMJD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human JMJD4. Alternate names: FLJ12517, MGC129896ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57088