Learn More
Aviva Systems Biology Corp JMJD4 Antibody - C-terminal region (ARP39573_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39573_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JMJD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human JMJD4. Alternate names: ApplicationWB. Species reactivity: Human, Mouse, Pig, Rat
Catalog No. 50-014-57089