Learn More
Aviva Systems Biology Corp Jmjd8 Antibody - C-terminal region (ARP58211_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58211_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Jmjd8. It was validated on Western Blot. . Alternate names: 2610003J06RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-286-452