Learn More
TARGETMOL CHEMICALS INC JMS-17-2 10MG
SDP
Catalog No. 502576655
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JMS-17-2 is a potent and selective antagonist of CX3CR1( IC50 0.32 nM). purity: 98%
Catalog No. 50-257-6655 Supplier TARGETMOL CHEMICALS INC Supplier No. T584910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JMS-17-2 is a potent and selective antagonist of CX3CR1( IC50 0.32 nM). purity: 98%