Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JMS-17-2 | 1380392-05-1 | MFCD30489012 | 99.2% | 419.95 | C25H26ClN3O | 50MG
SDP

Catalog No. 5000235838
Encompass_Preferred

JMS-17-2 is a small-molecule research compound that functions as a potent and selective antagonist of the CX3CR1 receptor (IC50 = 0.32 nM). It has been used in preclinical studies to impair metastatic seeding and colonization of breast cancer cells. The material is supplied as a high-purity solid with recommended cold storage for prepared solutions to preserve stability.

  • Chemical formula: C25H26ClN3O.
  • Molecular weight: 419.95 g·mol-1.
  • High purity suitable for research use.
  • Appearance: white to off-white solid.
  • Storage: store solid at 4°C protected from light; store solutions at -20°C to -80°C for longer-term stability.
  • Applications: in vitro and in vivo studies of CX3CR1-mediated processes and metastasis models.
  • Available in multiple pack formats including small mg quantities and ready-to-use DMSO solutions.

Catalog No. 50-002-35838 Supplier Medchemexpress LLC Supplier No. HY12391850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JMS-17-2 is a small-molecule research compound that functions as a potent and selective antagonist of the CX3CR1 receptor (IC50 = 0.32 nM). It has been used in preclinical studies to impair metastatic seeding and colonization of breast cancer cells. The material is supplied as a high-purity solid with recommended cold storage for prepared solutions to preserve stability.

  • Chemical formula: C25H26ClN3O.
  • Molecular weight: 419.95 g·mol-1.
  • High purity suitable for research use.
  • Appearance: white to off-white solid.
  • Storage: store solid at 4°C protected from light; store solutions at -20°C to -80°C for longer-term stability.
  • Applications: in vitro and in vivo studies of CX3CR1-mediated processes and metastasis models.
  • Available in multiple pack formats including small mg quantities and ready-to-use DMSO solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.