Learn More
Medchemexpress LLC HY-123918 5mg Medchemexpress, JMS-17-2 CAS:1380392-05-1 Purity:>98%
SDP
Catalog No. 501969651
Encompass_Preferred
Medchemexpress, HY-123918 5mg JMS-17-2 CAS:1380392-05-1 JMS-17-2 is a potent and selective CX3CR1 antagonist with an IC50 of 0.32 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9651 Supplier Medchemexpress LLC Supplier No. HY1239185MG
Medchemexpress, HY-123918 5mg JMS-17-2 CAS:1380392-05-1 JMS-17-2 is a potent and selective CX3CR1 antagonist with an IC50 of 0.32 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.