Learn More
Medchemexpress LLC HY-U00433A 5mg Medchemexpress, JMV 2959 hydrochloride CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU00433A5MG
Encompass_Preferred
Medchemexpress, HY-U00433A 5mg JMV 2959 hydrochloride CAS: JMV 2959 hydrochloride is a growth hormone secretagogue receptor type 1a ( GHS-R 1a ) antagonist with an IC 50 of 32±3 nM in LLC-PK 1 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4445